Social Distance Slip-Gard™ 6×24" Floor Sign: Wait Here to Step Forward - Red, Accuform®
# de catalogue 76416-920
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Color:Red/White/Black
- Cat. no.:76416-920
- Legend:WAIT HERE UNTIL CALLED TO STEP FORWARD (Footprints)
Spécifications
A propos de cet article
Floor signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus.
- Constructed of thick adhesive vinyl
- Non-slip, pebble-textured surface provides added durability
- Strong, permanent, acrylic adhesive back with peel-off liner
- Adheres firmly to smooth, clean, dry surfaces
Subsurface is printed and laminated to protect the graphics and message from heavy pedestrian traffic.
Customers Who Bought This Also Bought