A propos de cet article
These stainless steel, auto-fill vapor phase cryogenic storage systems are designed to maintain temperatures of samples well below the critical −135 °C (−211 °F) range, ensuring long-term cell viability.
- Stainless steel fabrication
- Long-term cell viability ensured
- 4,8,12 13-2 mini racks
- 14,30,58 13-2 standard racks
Samples are stored in an isolated inner aluminum tank, free of liquid, limiting cross-contamination concerns associated with sample storage in liquid.
The rotating storage tank has a reduced-size access opening with an easy-to-handle hinged lid, which limits the induction of harmful warm air during lid openings and reduces the system's consumption of liquid nitrogen. Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up, controller settings and an employee inservice by a factory representative.
A fully automatic controller and alarm package allows for convenient liquid level maintance. Sample security is monitored with a full array of alarms, including low liquid level, high liquid level, lid open, sensor failure, and power failure. Fully operational 3 to 5 day battery backup provides added security. Systems also include foldable skid-free step, five lockable casters, and one 1.8 m (6') armor-plated transfer hose.
Spécifications
- Catalog No:
- 76411-330
- 76411-328
- 76411-324
- 76411-326
- 76411-320
- 76411-322
- 76411-316
- 76411-318
- 76411-314
- Model:
- AF-VPSP 3 PS
- AF-VPSP 3 CS
- AF-VPSP 2 PS
- AF-VPSP 3
- AF-VPSP 2
- AF-VPSP 2 CS
- AF-VPSP 1 CS
- AF-VPSP 1 PS
- AF-VPSP 1
- Evaporation rate:
- 15 L/Day
- 15 L/Day
- 9 L/Day
- 15 L/Day
- 9 L/Day
- 9 L/Day
- 8 L/Day
- 8 L/Day
- 8 L/Day
- Ext.Ø:
- 151.1 cm
- 151.1 cm
- 114.3 cm
- 151.1 cm
- 114.3 cm
- 114.3 cm
- 86.3 cm
- 86.3 cm
- 86.3 cm
- Ø int.:
- 139.7 cm
- 139.7 cm
- 102.9 cm
- 139.7 cm
- 102.9 cm
- 102.9 cm
- 75 cm
- 75 cm
- 75 cm
- Liquid nitrogen capacity:
- 150 L
- 150 L
- 76 L
- 150 L
- 76 L
- 76 L
- 46 L
- 46 L
- 46 L
- Neck int.Ø:
- 62 cm
- 62 cm
- 47.7 cm
- 62 cm
- 45.7 cm
- 45.7 cm
- 33 cm
- 33 cm
- 33 cm
- Vial capacity:
- 79300×2 ml vials
- 79300×2 ml vials
- 41600×2 ml vials
- 79300×2 ml vials
- 41600×2 ml vials
- 41600×2 ml vials
- 19500×2 ml vials
- 19500×2 ml vials
- 19500×2 ml vials
- Weight Empty:
- 703 kg
- 703 kg
- 417 kg
- 703 kg
- 417 kg
- 417 kg
- 295 kg
- 295 kg
- 295 kg