VWR® CryoPro® Standard Auto-Fill Systems
Fournisseur: VWR International
Certificats
A propos de cet article
This Series offers all of the conveniences of an auto fill system in a full access cabinet design. This Series can easily be set to provide safe storage in vapor or liquid phase.
- Auto-fill convenience
- Large vial capacities
- Remote alarm contacts
- Digital temperature display
- Easy-to-select setting options
- VWR® two-year limited parts and labor and five-year limited vacuum warranty
Large capacity, auto-fill liquid nitrogen (LN₂) units in an economical steel and fiberglass cabinet.
A multicolored bar on the front panel provides easy setup and monitoring of liquid levels and level alarms.
The units feature alarms for high temperature, high and low liquid levels, and low LN₂ supply. The units are mounted on casters and have built-in lid locks. Exclusive sealed center shaft prevents hazardous spurting of LN₂ that can occur with a hollow shaft retrieval claw.
Ordering information: Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up (which includes controller settings, and personnel training by the manufacturer).
Delivery: Package systems include cryostorage unit, full inventory system (standard and mini storage racks, 100-cell and 25-cell boxes, and box dividers), 1.8 m (6') stainless steel transfer hose, liquid level measuring stick, and a pair of cryo-gloves.
Caution: Static evaporation rates and static holding times are nominal. Actual rates may be affected by the nature of the contents, atmospheric conditions, container history, and manufacturing tolerances.
Spécifications
- Catalog No:
- 76411-304
- 76411-306
- 76411-302
- 76411-312
- 76411-308
- 76411-310
- Model:
- AFIS-10CS
- AFIS-10PS
- AFIS-10
- AFIS-20PS
- AFIS-20
- AFIS-20CS
- Canister height:
- 533 mm (21") (Usable Internal Height)
- 533 mm (21") (Usable Internal Height)
- 533 mm (21") (Usable Internal Height)
- 533 mm (21") (Usable Internal Height)
- 533 mm (21") (Usable Internal Height)
- 533 mm (21") (Usable Internal Height)
- Ø int.:
- 533 mm
- 533 mm
- 533 mm
- 787 mm
- 787 mm
- 787 mm
- Static holding time (days):
- 33 days (nominal)
- 33 days (nominal)
- 33 days (nominal)
- 52 days (nominal)
- 52 days (nominal)
- 52 days (nominal)
- Vial capacity:
- 10400×2 ml vials
- 10400×2 ml vials
- 10400×2 ml vials
- 24050×2 ml vials
- 24050×2 ml vials
- 24050×2 ml vials
- Weight Full:
- 224 kg (537 lbs)
- 224 kg (537 lbs)
- 224 kg (537 lbs)
- 475.5 kg (1046 lbs)
- 475.5 kg (1046 lbs)
- 475.5 kg (1046 lbs)