Sanitizer Stand and Dispenser, Diversified Woodcrafts
Fournisseur: Diversified Woodcrafts
Certificats
A propos de cet article
Essential for today's public spaces, a complete hand sanitizer unit that includes the stand and motion-activated, touchless gel dispenser.
- Indoor/outdoor stand and dispenser
- Heavyweight, free-standing design
- Powercoated base
- Easy to assemble
- Universal mounting plate with theft-resistent screws or double-sided tape (not included)
- Touchless, motion-activated gel dispenser
- Dispenser includes a refillable plastic bottle (1000ml capacity)
- Requires 4 'C' batteries (not included)
- Stands 49.5" tall when assembled
The stand is suitable for indoor and outdoor use, with a durable powder-coated base and mounting plate to withstand the elements. It is easy to assemble and move. The dispenser mounts on the stand with a universal mounting plate that fits most dispensers. It can be mounted either with theft-resistant screws, or by two-sided tape (not included). The gel dispenser provides clean, accessible hand sanitization. It comes with a refillable, 1000ml capacity plastic bottle. Hand sanitizer is not included.
Ordering information: Available to purchase as a combo unit, stand only, or dispenser only. Batteries, double-sided tape, and hand sanitizer not included.
Packaging: Dispenser and stand are boxed separately.
Customers Who Bought This Also Bought