A propos de cet article
ENCORE 4250 and 4200 high-low, easy-access power tables offer smooth quiet power adjustment of back and height allowing staff to comfortably position patients for each exam or procedure promoting efficiency in each exam or procedure.
- Full Power adjustment of back and height by foot control, hand control available as an accessory
- Patient load 500 lbs.
- Pelvic tilt 7° and 11°
- Electrical outlets with circuit breaker - No fuses
- Instrument warming drawer, ON/OFF switch with integrated circuit breaker - No fuses
- Easy to adjust multi-position stirrups
- 4-year warranty
ENCORE 4250 High-Low Easy-Access table offers smooth quiet power adjustment of the back (flat to 80°) and height (24"- 40") to accommodate the majority of patient populations. Table features a 500 lb. patient capacity, dual electrical receptacles protected by an easy access circuit breaker (no fuses), an instrument warming drawer with the ON/OFF switch on the front side of the table for quick confirmation unit is active, independent operation between leg extension and a Stainless Steel irrigation basin, 7° and 11° pelvic tilt, multi-position stirrups, a field reversible side drawer, hand control port to allow an addition of the Hand Control accessory any time, seamless top to enhance infection control procedures and circuit breakers - NO FUSES for overload protection and 4 Year Warranty. Table ships complete with top securely attached to base of table - No assembly required.