Brady® Universal Plus Absorbent Pad, Brady Worldwide
# de catalogue 76417-804
Fournisseur: Brady Worldwide
Spécifications
- W×L:12×12"
- Color:Green
- Material:PP
- Absorbency:20 gal. (12.5 gl/gs)
Spécifications
A propos de cet article
Universal Plus products are chemically inert and green in color (the universal symbol for safety) to indicate that they're intended for use with hazardous materials.
- Surfactant-treated; brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
- Medium weight design is optimal for most general maintenance needs
- Universal Plus products feature 3-ply construction, increasing durability while reducing lint
Universal plus absorbent pads are non-perforated absorbents designed to offer the greatest absorbency and better tear-resistance in high traffic areas. These dimpled, medium-linting, and environmentally friendly pads are also fire-resistant and highly durable (15 to 27 lbs.).
Certifications: EPA 40 CFR 112.7(c)(1)(vii);OSHA 29 CFR 1910.120(j)(1)(vii);OSHA 29 CFR 1910.22(a)(2)
Customers Who Bought This Also Bought