ContactUsLinkComponent
Blue Phantom® Musculoskeletal Knee Ultrasound Trainer
Blue Phantom® Musculoskeletal Knee Ultrasound Trainer
# de catalogue 470327-402
MSK Knee Ultrasound Training Model
Blue Phantom® Musculoskeletal Knee Ultrasound Trainer
# de catalogue 470327-402
Spécifications
  • Description:
    Knee trainer
  • Cat. no.:
    470327-402
Spécifications
A propos de cet article
Constructed from a digital human file.
  • Superb image quality
  • Use with any ultrasound system
  • Self-healing tissue
  • Adjustable volume in the Bursa and Suprapatellar Space to simulate various injections and aspirations
  • Lightweight and easy to transport
Blue Phantom® is proud to introduce the World's first musculoskeletal ultrasound training model for joint injections of the knee. Constructed utilizing our innovative Life Cast™ modeling approach, the external landmarks were cast from a human knee while the internal anatomy is based on digital human files. This ultra-durable ultrasound simulator incorporates all of the anatomy required to teach, learn, and practice the skills associated with MSK joint injections and aspirations of the knee and was designed for both ultrasound guided and blind insertion procedural training. Utilizing advancements in SimulexUS™ technology we are able to accurately replicate the tendons and ligaments of the knee to offer a truly lifelike training environment, and because SimulexUS™ tissue is self-healing, the need to purchase replacement parts is dramatically reduced.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Customers Who Bought This Also Bought