Blue Phantom® PUBS Ultrasound Trainer
# de catalogue 470327-414
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Spécifications
- Description:PUBS trainer
- Cat. no.:470327-414
Spécifications
A propos de cet article
Superb image quality.
- Highly realistic
- Syringe actuated arterial pulsation
- Easy to refill
- Use with any ultrasound system
Blue Phantom®'s new percutaneous umbilical cord blood sampling (PUBS) - and Fetal Umbilical Blood Sampling (FUBS) ultrasound training model is an excellent tissue simulation trainer for ultrasound guided procedure training. Built as an extension on our amniocentesis ultrasound trainer, this hands-on training model contains an 18-week fetus with umbilical cord and offers the user an excellent skills development and assessment tool for teaching or learning the complete cordocentesis procedure.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Customers Who Bought This Also Bought