A propos de cet article
Excellent imaging characteristics.
- Anatomically correct adult torso
- Superb kidney architecture
- Ultra-durable self-healing tissue
- Matches the acoustic characteristics of real human tissue
- Use any ultrasound system
Blue Phantom® renal biopsy ultrasound training models provides users extremely realistic sonographic imaging characteristics and feels like real human tissue. Blue Phantom®'s patented ultra-durable self-healing tissue provides users with a low cost of ownership. In addition to training users how to utilize ultrasound to image the kidney, users are able to perform two types of procedures on the model core needle biopsy procedures and needle aspiration procedures.
Ultrasound imaging and procedural skills include; using ultrasound system controls, transducer positioning and movement, recognition of kidney anatomy, the appearance of ribs and avoiding them during renal biopsy, using ultrasound to target the appropriate location for renal biopsy, and performing an ultrasound guided renal biopsy procedure. This ultrasound hands-on training model will perform well using any ultrasound imaging system configured with the appropriate renal biopsy transducer.
This ultrasound phantom model is excellent for specialties including nephrology, interventional radiology, surgery, surgical training programs, ultrasound training programs, simulation centers, surgical skills centers, medical education facilities, renal biopsy device manufacturers, and ultrasound manufacturers for ultrasound education and demonstrations.