Blue Phantom® Lumbar Puncture Ultrasound Trainers
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
A propos de cet article
Available in a variety of configurations to meet your training needs.
- Extremely durable and self-healing
- Identify optimal insertion points, angle of needle insertion, and depth to the Ligamentum flavum, epidural space, and spinal cistern
- Superb ultrasound imaging characteristics
- Use any ultrasound system
- Use in upright or lateral position
Blue Phantom® Lumbar puncture and spinal epidural training model provides clinicians the flexibility to practice and teach a wide variety of procedures including lumbar puncture, lumbar epidural, thoracic epidural, and cervical epidural diagnostic and therapeutic procedures. This Lumbar puncture trainer with lumbar epidural is available in a variety of configurations to meet your training needs. The base model contains the lumbar puncture and lumbar epidural configuration. Users are able to add an additional spinal region to the base platform at an additional cost. These optional configurations include thoracic epidural and cervical epidural modules.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Customers Who Bought This Also Bought