A propos de cet article
Better training at a lower cost.
- Superb imaging characteristics
- Use with any ultrasound system
- Self-healing tissue offers tremendous durability
- No special storage needs
Blue Phantom®'s new pediatric ultrasound central line and regional anesthesia Training Model is the most realistic central line and nerve block simulator available anywhere. Using the innovative Life Cast™ modeling approach, the model provides the most realistic external landmarks while the internal anatomy is based on digital human files of a six-year-old child. This ultra-durable mannequin incorporates all the anatomy required to gain proficiency in using ultrasound for pediatric central line placement and peripheral nerve blocks.
This model is excellent for both ultrasound guided and blind insertion central line procedural training. The ultra-durable self-healing tissue withstands tremendous use, minimizing the need for replacement parts. Blue Phantom® allows clinicians to teach, practice, and learn central line and regional anesthesia procedures repeatedly without the need to worry about cost per use. Perform complete central line placements of the internal jugular and subclavian vein.