A propos de cet article
Compact and lightweight.
- Can be used with any ultrasound system
- Self-healing and ultra-durable tissue
- Simulate arterial pulsation via the included hand bulb
- Available in transparent or opaque material
The internal jugular central line ultrasound simulator offers superb internal and external anatomical realism. Developed using the innovative Life Cast™ modeling techniques, the external anatomy is presented in tremendous detail and the internal anatomy matches human anatomy, allowing you to practice and teach on this extremely lifelike central line manikin. The specially designed tissues will never clog needles and will always image like human tissue without unrealistic ultrasound system settings.
The lightweight and portable internal jugular training manikin contains vascular anatomy including the internal jugular vein (IJ) and the carotid artery. Internal structures also include the trachea and clavicle providing you with superb realism – using any ultrasound system equipped with a vascular access ultrasound transducer. External landmarks also include a palpable suprasternal notch, trachea, and clavicle. This central venous access ultrasound phantom is specifically designed for teaching and practicing internal jugular (IJ) cannulation; the training model accommodates both ultrasound guidance and blind insertion technique.