A propos de cet article
Superior realism, reliability, and durability.
- Self-healing tissue offers users tremendous durability
- Anatomically correct; constructed from a digital human file
- Veins are compressible while the arteries remain uncompressed
- Easy to refill vessels using Quick fill™ port
Blue Phantom® is proud to introduce the most realistic ultrasound central line mannequin available anywhere. Constructed utilizing the innovative Life Cast™ modeling approach, the external landmarks were cast from a live human while the internal anatomy is based on digital human files. This ultra-durable ultrasound simulator incorporates all of the anatomy required to teach, learn, and practice the skills associated with central line placement and was designed for both ultrasound guided and blind insertion procedural training.
The self-healing tissue withstands tremendous use, minimizing the need for replacement parts. Blue Phantom®'s new upper torso central venous catheter model offers clinicians the most life-like training environment at a low cost of ownership. Perform complete central line placements of the internal jugular and subclavian vein - including needles, guidewires, dilation, and threading of catheters. On all models arterial pulsations are provided using a hand pump, or an integrated automatic pump.