Blue Phantom® Transvaginal Ultrasound Trainers
Fournisseur: Avantor
Certificats
A propos de cet article
Specific training for transvaginal ultrasound.
- Five unique trainers
- Includes storage case and privacy skirt
- Extremely realistic imaging characteristics
- Feels like real human tissue
- Anatomically accurate external anatomy
With accurate external anatomy and various internal structures, these ultrasound trainers provide accurate imaging with clear diagnostic structures. Each trainer is an individual pelvic model shipped with a padded case. General pathology model:
Uterus with prominent endometrium, left and right ovaries, periovarian abscess, broad ligaments, ovarian follicles, free fluid in the Cul-de-Sac, bowel, bladder, and other accessory pelvic structures.
Sonohysterography and sonosalpingography model:
cervix, cervical canal, uterus, endometrial canal, fibroids, bladder, patent fallopian tubes, and bowel.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Customers Who Bought This Also Bought