STEAM Cart Mobile Learning Platforms, BioFit Engineered Products
Fournisseur: BioFit
Certificats
A propos de cet article
Used to transport resources between classrooms or as a standalone multipurpose activity center, the BioFit STEAM Cart is a versatile mobile learning platform specially designed to help educate tomorrow’s innovators; its rugged, kid-friendly construction and flexibility make it a popular solution in today’s most progressive K-12 learning environments and maker spaces alike.
- Two, large 4" deep trays provide plenty of space and keep gear secured during transport
- Twin side-panel pegboards for holding tools, supplies and more
- All-steel welded construction tested and proven in tough industrial settings
- Dual-wheel casters rated at 100 lbs. each for a total cart capacity of 400 lbs.
- Durable powder-coat finish in six standard colors stands up to years of use
- Nimble, smooth and ready to roll, with a compact profile ideal for confined spaces
- 13 year warranty helps schools of any size to maximize their bottom lines
The BioFit STEAM cart is brought to you by the company with over 75 years experience delivering creative and reliable ergonomic seating solutions, cafeteria tables, book trucks and more to the education market place.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Spécifications
- Catalog No:
- 76415-424
- 76415-422
- 76415-420
- 76415-430
- 76415-428
- 76415-426
- Width:
- 28"
- 28"
- 28"
- 28"
- 28"
- 28"
- Depth:
- 17"
- 17"
- 17"
- 17"
- 17"
- 17"
- Height:
- 34⅝"
- 34⅝"
- 34⅝"
- 34⅝"
- 34⅝"
- 34⅝"
- Weight:
- 31 lbs.
- 31 lbs.
- 31 lbs.
- 31 lbs.
- 31 lbs.
- 31 lbs.