ContactUsLinkComponent
Test Strips, Chloride, Dosatest, VWR Chemicals BDH®
Test Strips, Chloride, Dosatest, VWR Chemicals BDH®
# de catalogue BDH85441.601
Fournisseur:  VWR International
Numéro CAS:  
Vwr* chloride test strip, for the semi-quantitative determination of chloride (Cl - ) in solutions, Gradation of scale: 0-500-1000-1500-2000-3000 mg/l Cl - brown to yellow, In the pH range 2 to 12 the reaction is independent of the pH value of the solution to be analysed, 500 dosatest
Test Strips, Chloride, Dosatest, VWR Chemicals BDH®
# de catalogue BDH85441.601
Fournisseur:  VWR International
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chloride test 0 - 3000 mg/l
  • Range:
    0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/l
  • No. of Strips:
    100
Spécifications
A propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Ideal to control your disinfection. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6 x 95 mm.
Customers Who Bought This Also Bought