Avantor® ACE® Excel® Oligo, Analytical, HPLC/UHPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® Oligo is a unique encapsulated stationary phase for high resolution ion-pair reversed-phase analysis of oligonucleotides. These high pH stable stainless steel columns are compatible with all buffers and mobile phases typically used for oligonucletide analysis by LC-UV and LC-MS, including TEAA and HFIP.
- Ultra-inert, base deactivated silica for exceptional reproducibility and peak shape
- Excellent column lifetime under oligonucleotide testing conditions
- Ultra-low bleed for excellent LC/MS compatibility
High efficiency 1.7 and 3.0 µm particle sizes for high resolution oligonucleotide separations.
These columns are made of stainless steel.
Spécifications
- Catalog No:
- 76415-438
- 76415-436
- 76415-434
- 76415-446
- 76415-444
- 76415-442
- 76415-440
- 76415-432
- 76415-450
- 76415-448
- Application:
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Oligonucleotides
- Carbon load:
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- pH range:
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g