Prism
# de catalogue 470092-284
Fournisseur: United Scientific Supplies
Spécifications
- Description:Prism, Equilateral/Acrylic, 50×25 mm
- Cat. no.:470092-284
- Supplier No.:PAE050
Spécifications
A propos de cet article
Equilateral prisms, made from high quality clear acrylic sheet.
- Highly polished surfaces
- 50×25 mm
This prism allows you to investigate the phenomena of refraction and internal reflection.
Customers Who Bought This Also Bought