Tingley® Boot Saver® Disposable Latex Shoe Covers, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Place over boots or shoes for 100% liquid-proof protection against chemicals, oils, and water.
- Durable, natural rubber latex stretches over shoes or boots
- Withstands snags, tears, and abrasions
- Textured outsole, non-slip shoe cover provides grip on wet and dry surfaces
- Ambidextrous design allows boot covers to fit both right and left feet
- Powder-free finish eliminates concerns of powder contamination
Latex shoe covers are resistant to most natural oils, water-soluble compounds, chemicals, low concentration acids and alkalis, alcohols, dilute bleach, and mild to moderate strength detergents and cleaners. Ideal for food processing plants, maintenance and service technicians, paint spray booths, laboratories, chemicals, and more.
Spécifications
- Catalog No:
- 76415-034
- 76415-056
- 76415-032
- 76415-054
- 76415-042
- 76415-038
- 76415-040
- 76415-026
- 76415-048
- 76415-028
- 76415-050
- 76415-046
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial