Transparent Tapes, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
High-quality porous clear tape is perforated for easy bi-directional tearing.
- Patient specific, single use rolls
- Contributes to the safe and sanitary treatment of all patients
- Conforms and stretches to accommodate movement
- No natural rubber latex
Transparent tape is excellent for I.V. sites, dressings, and tubing retention.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.