Paper Tapes, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Microporous paper tape is lightweight, gentle on skin, won't harm delicate wounds, and will hold even when wet.
- Patient specific, single use rolls
- Contributes to the safe and sanitary treatment of all patients
- No natural rubber latex
Paper tape is recommended for use on delicate and elderly skin.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.