A propos de cet article
The compact MINIPULS® 3 is a high-performance, low-pulse, peristaltic pump with interchangeable pump heads for delivering a diverse variety of liquids from biological to chemical without altering the critical components within the liquid.
- Pulse-free liquid transfer
- Flow rates to meet your needs
- Easy to operate
- Six different channel and flow rate configurations are available
Utilizing positive displacement pump technology, liquid flows nearly pulse-free as a result of as many as ten stainless steel rollers. The MINIPULS 3 produces smoother flows with excellent accuracy and precision, even at high flow rates. Its adjustable speed stepper motor and high-quality precision bore tubing assure reproducible flow rates.
With a choice of six quickly interchangeable pump heads, a wide range of flow rates is available to use for various applications.
Four standard models (1, 2, 4, or 8 channels): 0.3 to 30 ml/min.
Two high flow models (2 or 4 channels): 1 to 220 ml/min.
MINIPULS 3 is easy to install, simple to operate, and easy to maintain. With an uncomplicated keypad control and easy-to-read LCD screen, access functions for start/stop, flow direction, increase/decrease flow rate, and prime. Remote control is also possible.