Type VII X-Ray Detectable Gauze, DUKAL™ Corporation
Fournisseur: Avantor
Certificats
A propos de cet article
Blue filament runs through each sponge and has a unique wave pattern to ensure easy X-ray detection.
- USP Type VII gauze, 100% woven-cotton
- Sterile sponges are packaged in rigid plastic tray with peel back lid
- No natural rubber latex
Both sterile and non-sterile sponges are banded in 10’s to ensure accurate count.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.