Type VII Tray Gauze, DUKAL™ Corporation
Fournisseur: Avantor
Certificats
A propos de cet article
USP Type VII gauze, 100% woven-cotton.
- Sponges are banded in 10’s to ensure accurate count
- No natural rubber latex
Packaged in rigid plastic tray with peel back lid.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.