American White Cross First Aid® Blue Metal Detectable Adhesive Strips, DUKAL™ Corporation
Supplier: Dukal
Certificates
About this item
The metal under the pad allows for dependable detection.
- Designed for use in the food processing industry
- The blue color allows the bandage to be easily spotted
These bandages are easy to open and have a highly absorbent, non-stick wound pad.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.