Sterile Post-Op Sponges, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
A multi-layer, woven gauze-faced sponge with cellulose fill.
- Well suited for use in place of a single traditional sponge
- No natural rubber latex
Post-Op sponges provide fast wicking and increased absorbency.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.