Retention Tapes, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Highly conformable, adhesive non-woven fabric tape.
- Printed s-curve release liner
- Pre-cut to eliminate waste
- Provides light compression without constriction
- No natural rubber latex
Tape is designed to secure primary dressings, catheters, and drainage tubes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.