Sterile Wound Closure Strips, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Wound closure strips provide support following early suture/staple removal.
- Keeps wound closed without significant tension, edema, stretch, or movement
- Translucent and porous
- No natural rubber latex
Adhesive backed, thin, non-woven, reinforced fibers are arranged in a bilateral pattern for high tensile strength.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.