Heratherm™ General Protocol Microbiological Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These general protocol incubators are designed with sample safety in mind, for applications in pharmaceutical, medical, food and research laboratories.
- Gravity convection provides gentle air flow and minimal drying out
- Corrosion-resistant stainless-steel chamber (AISI 430)
- Intuitive user interface for setting temperature
- Large, easy to read vacuum fluorescent display
- Exceptionally small footprint
- Nema 5-15 US plug type
The internal glass door allows sample viewing without impacting temperature.
Specifications
- Catalog No:
- 76411-573
- 76411-577
- 76411-628
- 76411-632
- 76411-636
- Temperature range:
- Ambient +5…75 °C
- Ambient +5…75 °C
- Ambient +5…75 °C
- Ambient +5…75 °C
- Ambient +5…75 °C
- Electrical:
- 120 V, 60 Hz
- 120 V, 60 Hz
- 120 V, 60 Hz
- 120 V, 60 Hz
- 120 V, 60 Hz