VWR® P2 Balances with Calibration Certificate
Supplier: VWR International
Certificates
About this item
VWR® P2-Series/C balances which include an ISO/IEC 17025:2017 Calibration Certificate are the answer for an organizations regulatory and compliance needs.
- Complete calibration data reported
- Internal, rechargeable battery
- Backlit, LCD screen
- External and internal calibration options
- RS232 and USB (A and B) communication interfaces
- Five year warranty
The economical balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These compact, portable balances offer a variety of standard features, such as a multiple weighing modes, advanced communication features, multiple weighing units, and quick stabilization time.
The P2 series features USB (A and B) and RS232 interfaces. The VWR-30000P2.H and VWR-60000P2.H models only feature a RS232 connection.
Certifications: ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate; CE marked
Ordering information: All units supplied with switching AC adapter, weighing pan, user manual, and quick start guide. Includes five-year warranty. Accessories not included.
Specifications
- Catalog No:
- 76378-364
- 76378-366
- 76378-362
- 76378-372
- 76480-262
- 76378-374
- 76480-264
- 76378-368
- 76480-266
- 76378-370
- 76480-268
- 76480-270
- Operating temperature range:
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- 15…30 °C (59…86 °F)
- Communication interface:
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- RS232, USB (A and B)
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V