Metro Indicator Flag for WavDri™, Drying Tray, Intermetro Industries
# de catalogue CA76407-174
Fournisseur: Metropolitan Wire
Spécifications
- Color:Red/White
- Description:Indicator flag
- Material:Polymer
- Cat. no.:CA76407-174
- Dimensions:3.625×0.156×0.75"
Spécifications
A propos de cet article
Quickly identify if WavDri™ trays have been rotated.
Visually alternates from red to white when using the clamshell rotation feature for a consistent and documented rotation process.
Customers Who Bought This Also Bought