Avantor® Hichrom, Needle Port for HPLC Systems
# de catalogue 76399-156
Fournisseur: HICHROM LIMITED
Avantor® Hichrom, Needle Port for HPLC Systems
# de catalogue 76399-156
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Needle port
- Material:PEEK
- Cat. no.:76399-156
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
50,14 $ to 501,54 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
57,66 $ to 576,72 $
Cap, 12mm solid top storage, 13-425 Closure screw Thread, Color: Black, Material: Polypropylene
Fournisseur: Thermo Scientific
171,81 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $