Avantor® Hichrom, Frit Assembly
# de catalogue 76399-148
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Frit assembly, 0.25×1.5 mm
- Material:PEEK
- Cat. no.:76399-148
Spécifications
A propos de cet article
The frit assembly is made of PEEK.
Customers Who Bought This Also Bought
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$461.12 to $461.12
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$205.86
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$180.74
WHEATON® MicroLiter Crimp-Top Vials, 11 mm,Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$142.31
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$438.81
Preassembled Vial Kits, 8-425, Screw-Thread, Unmarked Vials, Septa and Caps, Restek
Fournisseur: Restek
$764.77