Spécifications
- Description:MakeBlock mBot-S explorer kits
- Cat. no.:470324-122
Spécifications
A propos de cet article
Entry-level educational robot for STEAM education and targeting students.
- Contains many electronic modules
- Updated version of mBot
- Comes with an LED matrix display
- Ages 8+
The MakeBlock mBot-S explorer kit is an entry-level educational robot for STEAM education and suitable for students from ages eight and above. It contains many electronic modules such as a LED matrix, sensors, an IR transmitter, and IR receiver. It has a companion programming software mBlock (for PC/mobile) and supports the Makeblock app for 3D construction.
mBot-S is an updated version of mBot. In addition to the features that mBot provides, mBot-S also comes with an LED matrix display. Students can put the LED display in any position as they please: a front, back, at the top. the LED matrix display brings diversity into mBot-S, allowing students to customize images by programming the RBG lights. Students turn the LED display into a board showing scores, weather forecasts, or emotions.