Cutting Mat
# de catalogue 470318-258
Fournisseur: KIDDER MFG. CO. LTD
Spécifications
- Description:Cutting mat
- L×W:60×45 cm
- Cat. no.:470318-258
Spécifications
A propos de cet article
Cut safely.
- Grid, 1 cm
Mat used as a surface for cutting materials or objects with a fine blade.
Customers Who Bought This Also Bought