MSA Mannitol Salt Agar Plates
# de catalogue 470192-610
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Pack of 10
- Description:MSA plates
- Cat. no.:470192-610
- Supplier No.:880992
Spécifications
A propos de cet article
Prepared media plates.
- Microbiology
We recommend purchasing prepared media plates no more than two weeks before use for best results, as these items are highly perishable.
Customers Who Bought This Also Bought