VWR® Concept Carts | Avantor
VWR® Concept Carts
Concept cart with phenolic top, light neutral, 330 lbs., Base: 27.56D×19.69"W, 27.17D×15.75”W (integrated bottom shelf), 19.69", 25.59 - 35.43"
Catalog # 76383-288
Supplier:   VWR International
VWR® Concept Carts
Catalog # 76383-288
Supplier:  VWR International
VWR® Concept Carts
VWR® Concept Carts
Concept cart with phenolic top, light neutral, 330 lbs., Base: 27.56D×19.69"W, 27.17D×15.75”W (integrated bottom shelf), 19.69", 25.59 - 35.43"
Price
$2,055.06
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Concept cart is a steel-framed additional work surface with height adjustment; suitable for many purposes.
  • Light neutral or snow white, powder-coated steel frame
  • Phenolic top
  • Integrated bottom shelf
  • Height adjustment is 25.59 to 35.43" with an allen key
  • Four swivel castors (Ø 3.94"), two with brakes
  • An extra shelf is available as an accessory
The integrated bottom shelf offers much needed extra storage space and an additional shelf is available as an accessory. Easy and effortless to move around, but with castors with brakes the cart stays firmly in place if needed. Well suited for use with all the workbenches.
Ordering information: Bottom shelf is included. Middle shelf is sold separately.
Delivery information: Product ships knocked-down.
Packaging: Flat-packed in cardboard box.
Spécifications
  • Largeur:
    19.69"
  • Désignation:
    Concept cart with phenolic top, light neutral
  • Dimensions:
    Base: 27.56D×19.69"W
  • Hauteur:
    25.59 - 35.43"
  • Shelf Dimensions:
    27.17D×15.75”W (integrated bottom shelf)
  • Load Capacity:
    330 lbs.
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Middle shelf for Concept cart, light neutral, 55 lbs. load capacity, 19.69D×15.75"W
    Middle shelf for Concept cart, light neutral, 55 lbs. load capacity, 19.69D×15.75"W
    76383-290
    Log in pour la disponibilité
    Each - $127.00