VWR® Shaker Flasks
Supplier: VWR International
Certificates
About this item
VWR® 5 L shaker flasks are perfect for shaker culture applications and storage.
- Plug seal and vent caps are available
- Vent caps with 0.2 µm hydrophobic filters for gas exchange without contamination
- Polycarbonate bottle has a high standard of transparency, clear scale and allows easy observation
- Graduated markings
- Packaged individually and sterilized by E-Beam
- Non-pyrogenic
Ideal for shaker culture applications, the recommended volume is 30 to 40% of the bottle volume.
Customers Who Bought This Also Bought