ContactUsLinkComponent
pH Electrode Saver Bottles, Cole-Parmer®
pH Electrode Saver Bottles, Cole-Parmer®
Catalog # 76377-400
Supplier:  Argos Technologies
Electrode Storage Bottle Large 10 Ml
pH Electrode Saver Bottles, Cole-Parmer®
Catalog # 76377-400
Supplier:  Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Electrode storage bottle, 50 ml, 4×1.5" (H×Ø)
  • Cat. no.:
    76377-400
Specifications
About this item
These storage bottles keep electrodes in working order.
  • Protect against bumps, spills, and drying out
  • Accepts electrodes up to Ø 13 mm
Large bottle includes one solid cap to reduce evaporation of storage solution and one O-ring cap for use with electrodes. Small bottle includes one O-ring cap and is a cost-effective option when storage solution is discarded and not regularly re-used.
Customers Who Bought This Also Bought