xtra TT Heavy-Duty Ultrasonic Cleaners, Elma
Supplier: Elma
Certificates
About this item
Robust benchtop Elmasonic xtra TT industrial ultrasonic cleaners are designed for operations that run continuously for up to 8 hours/day.
- Durable cleaners for production environment
- Sweep mode for uniform cleaning
- Dynamic mode for extra cleaning power
- Timer, ball valve drain, and heater 30 to 80 ⁰C
Certifications: CE, ISO9001, ISO13485.
Delivery: All models supplied with covers. Baskets and other accessories must be ordered separately.
Specifications
- Catalog No:
- 76326-630
- 76326-674
- 76326-678
- 76326-684
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health, Waste Reducing
- Sustainable Packaging, Safer Human and Environmental Health, Waste Reducing
- Sustainable Packaging, Safer Human and Environmental Health, Waste Reducing
- Sustainable Packaging, Safer Human and Environmental Health, Waste Reducing