Single-Use High Pressure Transfer Hose with SterilEnz™ Connections
Supplier: AVANTOR FLUID HANDLING, LLC
Certificates
About this item
Avantor's high-pressure single-use transfer hose with Avantor SterilEnz™ connections maintain the durability of high-pressure hose products while providing the benefits and flexibility of single-use assemblies.
- Suitable for elevated temperatures and pressures up to 1740.4 psi
- 1" inner diameter for high flow rate applications
- Gamma-irradiated with SAL 10⁻⁶
- Offered at 5 and 10 foot lengths
- Gasket affixed to TC face even when disconnected to prevent misalignment
Designed for applications where pressures would exceed normal biopharmaceutical processes.
Avantor single-use transfer hose with SterilEnz™ connections are pre-sterilized; then packaged in a double polybag as an added convenience for customers when working with the product. A 1" inner diameter supports high-flow rates, while the 5 and 10 foot lengths meet the requirements for many bioprocessing applications, with custom lengths available upon request.
Certifications: USP Class VI, ACDF, Sterile, Pressure rating up to 1740.4 psi.
Customers Who Bought This Also Bought