Forceps, Gemologist, Starlight Coated Tip, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
Designed for handling gem stones.
- Starlight coated tips hold gems more securely
- Various sizes available
- Stainless steel corrosion resistant
These gemologist straight medium point tweezers are ideal for handling medium gems. The handles are serrated for better control. They feature 'starlight' coated tips.
Customers Who Bought This Also Bought