Mini Spatula with Egonomic Handle, Excelta®
Supplier: Excelta
Certificates
About this item
Ergonomic mini spatulas (oilers) feature a soft, cushioned, and ESD safe (10¹⁰ ohms/sq.) handle.
- Overall length of each spatula is 76.2 mm (3")
Mini spatulas are ideal for placing small amounts of epoxy or lubricant into tight places.
Customers Who Bought This Also Bought