Scalpel handles, Excelta Corp®
Fournisseur: Avantor
Certificats
A propos de cet article
These scalpel handles and blades with the exception of the 181-22, are made of AISI 440 stainless steel and are cleanroom safe.
Customers Who Bought This Also Bought