ContactUsLinkComponent
Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
# de catalogue 76333-462
Fournisseur:  Chemglass
Lock Male Luer Swabable Pc Silicone PK25
Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
# de catalogue 76333-462
Fournisseur:  Chemglass
Spécifications
  • Description:
    Male Luer Lock, needleless injection site, swabable, polycarbonate, silicone
  • Cat. no.:
    76333-462
Spécifications
A propos de cet article
Male Luer lock with needleless injection site and swabable polycarbonate, silicone with lach samplers for bioreactors.
Customers Who Bought This Also Bought