Level Sensor Adapters, for Bioreactors, Chemglass
# de catalogue 76333-454
Fournisseur: Chemglass
Spécifications
- Description:M10 adapter to 1/4" for Level sensors
- Cat. no.:76333-454
Spécifications
A propos de cet article
Used to install a ¹/₄” OØ (6.35 mm) level sensor into an M10 headplate port.
Customers Who Bought This Also Bought