Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
# de catalogue 76333-450
Fournisseur: Chemglass
Spécifications
- Description:Large diameter sampling tube/harvest tube, m18, for 2-5 Liter bioreactors
- Cat. no.:76333-450
Spécifications
A propos de cet article
Pipe will fit a 2 to 5 liter reactor and has a lower diameter of ¹/₂" and a length of 212 mm into the reactor. The serrated hose connection is also ¹/₂" (12.7 mm) OØ. Can also be used as a quick way to harvest material due to its size.
Customers Who Bought This Also Bought