Stoppers/Adapters, Extraction Nut, 3/4" NPT, Chemglass
# de catalogue 76333-406
Fournisseur: Chemglass
Spécifications
- Description:45/50 ptfe stopper, 3/4" npt, complete with extraction nut
- Cat. no.:76333-406
Spécifications
A propos de cet article
PTFE stopper adapter is similar to CG-3002-L but with the addition of one female NPT port in the center.
The optional PTFE compression fittings provide a convenient way to insert probes or connect PFA tubing or FEP tubing to reactors or flasks. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.
Customers Who Bought This Also Bought