Offset Adapter, for Velp Hotplates, Chemglass
# de catalogue 76333-664
Fournisseur: Chemglass
Spécifications
- Description:Offset adapter, for velp hotplates, stainless steel, supplied with m8 bolt
- Cat. no.:76333-664
Spécifications
A propos de cet article
Offset Adapter, for Velp Hotplates, Stainless steel
This stainless steel offset adapter allows use of the CG-1995-V-22 vertical support rod with larger diameter accessories on CG-1994-V, CG-1995-V, and CG-1997-V series hotplates. Offset adapter can be used with cooling/heating blocks up approx. 8 to 5/8"/220 mm diameter. Offset created by adapter is 20 mm center to center.
Delivery: Supplied complete with M8 stainless steel bolt.
Customers Who Bought This Also Bought