Thermoregulator, Digital, Built in Timer with Clamp for mounting on Vertical Support Rod, Velp
# de catalogue 76333-658
Fournisseur: Chemglass
Spécifications
- Description:Digital thermoregulator only, with built in timer
- Cat. no.:76333-658
Spécifications
A propos de cet article
Digital thermoregulator is equipped with innovative Fuzzy Logic technology that provides accurate set point ±0.5 °C. Digital count down timer shuts off heating- adjustable from 0 to 24:59 (h:min). Innovative design incorporated a clamp for mounting on the vertical support rod.
Customers Who Bought This Also Bought